1-Chloro-4-fluoro-2-nitrobenzene ≥98.0%
Supplier: TCI America
C2166-5G
TCC2166-5GEA
204.29
USD
TCC2166-5G
1-Chloro-4-fluoro-2-nitrobenzene ≥98.0%
1-Chloro-4-fluoro-2-nitrobenzene
CAS Number: 345-17-5
MDL Number: MFCD00084853
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 239
Melting point (°C): 39
Flash Point (°C): 109
Formula:
C₆H₃ClFNO₂ MW: 175.55 g/mol Boiling Pt: 239 °C Melting Pt: 39 °C Flash Pt: 109 °C |
MDL Number:
MFCD00084853 CAS Number: 345-17-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...