Carbon nanotube multi-walled 20-40nm (diam.), 1-2µm (length)
Supplier: TCI America
Synonyms:
CNT
C2155-1G
TCC2155-1GEA
95.41
USD
TCC2155-1G
Carbon nanotube multi-walled 20-40nm (diam.), 1-2µm (length)
Carbon nanotube
CAS Number: 308068-56-6
Form: Powder
Color: Black
Other Product Names / Synonyms: Multi-walled Carbon Nanotube
Formula:
C MW: 12.011 g/mol |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...