TCIAC2152

C2152-1G
TCC2152-1GEA 85.36 USD
TCC2152-1G
TCIAC2152
CAS Number: 308068-56-6
Form: Powder
Color: Black

Other Product Names / Synonyms: Multi-walled Carbon Nanotube
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR