Carbon nanotube multiwalled below 10nm (diam.), 5-15µm (length)

Supplier: TCI America

C2148-1G
TCC2148-1GEA 130.45 USD InStock
TCC2148-1G
Carbon nanotube multiwalled below 10nm (diam.), 5-15µm (length)
Carbon nanotube multiwalled

CAS Number: 308068-56-6
Form: Powder
Color: Black


Other Product Names / Synonyms: Multi-walled Carbon Nanotube

Formula: C
MW: 12,011 g/mol
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR