Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon nanotube bundled multiwalled , Length 5-15 µm, Diameter <10 nm

Supplier: TCI America


C2147-200MG
TCC2147-200MGEA 70.78 USD InStock
TCC2147-200MG
Carbon nanotube bundled multiwalled , Length 5-15 µm, Diameter <10 nm
Carbon nanotube bundled multiwalled
Allowable temperature limit : 620 °C
Form: Powder
Color: Black


CAS Number: 308068-56-6
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty