TCIAC2146

C2146-200MG
TCC2146-200MGEA 214.43 USD
TCC2146-200MG
TCIAC2146
CAS Number: 308068-56-6
Form: Powder
Color: Black

Other Product Names / Synonyms: Herringbone Carbon Nanotube
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR