TCIAC2143

C2143-200MG
TCC2143-200MGEA 188.4 USD
TCC2143-200MG
TCIAC2143
CAS Number: 308068-56-6
Form: Powder
Color: Black

Other Product Names / Synonyms: Double-walled Carbon Nanotube
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR