5-Chloro-2-adamantanone ≥98.0%
Supplier: TCI America
Synonyms:
5-Chloroadamantan-2-one
C2140-1G
TCC2140-1GEA
90
USD
TCC2140-1G
5-Chloro-2-adamantanone ≥98.0%
5-Chloro-2-adamantanone
CAS Number: 20098-17-3
MDL Number: MFCD00798599
Molecular Formula: C10H13ClO
Molecular Weight: 184.66
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 200
Formula:
C₁₀H₁₃ClO MW: 184.66 g/mol Melting Pt: 200 °C |
MDL Number:
MFCD00798599 CAS Number: 20098-17-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...