4-Chloro-2-fluorobenzonitrile ≥98.0%
Supplier: TCI America
C2121-25G
C2121-5G
TCC2121-5GEA
75
USD
TCC2121-5G
TCC2121-25G
4-Chloro-2-fluorobenzonitrile ≥98.0%
4-Chloro-2-fluorobenzonitrile
CAS Number: 57381-51-8
MDL Number: MFCD00143284
Molecular Formula: C7H3ClFN
Molecular Weight: 155.56
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 60
Formula:
C₇H₃ClFN MW: 155.56 g/mol Melting Pt: 60 °C |
MDL Number:
MFCD00143284 CAS Number: 57381-51-8 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...