4-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Chloro-3-methylbenzoic acid
C2115-5G
TCC2115-5GEA
16.24
USD
TCC2115-5G
4-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)
4-Chloro-m-toluic acid
CAS Number: 7697-29-2
MDL Number: MFCD00045853
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 213
Other Product Names / Synonyms: 4-Chloro-m-toluic Acid
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol Melting Pt: 213 °C |
MDL Number:
MFCD00045853 CAS Number: 7697-29-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...