6-Chlorouracil ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-(3-Bromophenyl)pyrazole
, 6-Chloro-2,4(1H,3H)-pyrimidinedione
, 6-Chloro-2,4-dihydroxypyrimidine
, 4-Chlorouracil
, 4-Chloro-2,6-dihydroxypyrimidine
, 6-Chloropyrimidine-2,4(1H,3H)-dione
C2093-25G
C2093-5G
TCC2093-25GEA
66.99
USD
TCC2093-25G
TCC2093-5G
6-Chlorouracil ≥98.0% (by titrimetric analysis)
6-Chlorouracil
CAS Number: 4270-27-3
MDL Number: MFCD00014595
Molecular Formula: C4H3ClN2O2
Molecular Weight: 146.53
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 295
Formula:
C₄H₃ClN₂O₂ MW: 146.53 g/mol Melting Pt: 295 °C |
MDL Number:
MFCD00014595 CAS Number: 4270-27-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...