6-Chloronicotinonitrile ≥97.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
2-Chloropyridine-5-carbonitrile
, 6-Chloro-3-pyridinecarbonitrile
, 2-Chloro-5-cyanopyridine
C2056-5G
C2056-25G
TCC2056-5GEA
50.75
USD
TCC2056-5G
TCC2056-25G
6-Chloronicotinonitrile ≥97.0% (by GC, total nitrogen)
6-Chloronicotinonitrile
CAS Number: 33252-28-7
MDL Number: MFCD00084941
Molecular Formula: C6H3ClN2
Molecular Weight: 138.55
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 107
Melting point (°C): 117
Other Product Names / Synonyms: 6-Chloronicotinonitrile
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol Boiling Pt: 107 °C Melting Pt: 117 °C |
MDL Number:
MFCD00084941 CAS Number: 33252-28-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...