(8-Chloro-1-naphthylthio)acetic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
C2052-5G
TCC2052-5GEA
36.52
USD
TCC2052-5G
(8-Chloro-1-naphthylthio)acetic acid ≥95.0% (by titrimetric analysis)
(8-Chloro-1-naphthylthio)acetic acid
CAS Number: 129-94-2
MDL Number: MFCD00043901
Molecular Formula: C12H9ClO2S
Molecular Weight: 252.71
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 157
Formula:
C₁₂H₉ClO₂S MW: 252.71 g/mol Melting Pt: 157 °C |
MDL Number:
MFCD00043901 CAS Number: 129-94-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...