2-Chloro-6-fluorophenol ≥98.0%
Supplier: TCI America
C2049-5G
C2049-25G
TCC2049-5GEA
32.48
USD
TCC2049-5G
TCC2049-25G
2-Chloro-6-fluorophenol ≥98.0%
2-Chloro-6-fluorophenol
CAS Number: 2040-90-6
Molecular Formula: C6H4ClFO
Molecular Weight: 146.55
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 66
Formula:
C₆H₄ClFO MW: 146.55 g/mol Melting Pt: 66 °C |
MDL Number:
MFCD01631574 CAS Number: 2040-90-6 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...