Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Chlorovalerophenone ≥96.0%

Supplier: TCI America
Synonyms: 1-(4-Chlorophenyl)-1-pentanone

C2041-25G
TCC2041-25G
4'-Chlorovalerophenone ≥96.0%
CAS Number: 25017-08-7
MDL Number: MFCD00018716
Molecular Formula: C11H13ClO
Molecular Weight: 196.67
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Boiling point (°C): 156
Freezing point (°C): 30

Other Product Names / Synonyms: 4'-Chloropentanophenone


Formula: C₁₁H₁₃ClO
MW: 196,68 g/mol
Boiling Pt: 100 °C (0.5 torr)
Melting Pt: 31 °C
MDL Number: MFCD00018716
CAS Number: 25017-08-7
EINECS: 246-566-4
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)
Freezing point (°C) 30
Boiling point (°C) 156


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty