Cyclohexylurea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 1-Cyclohexylurea

C2034-25G
TCC2034-25GEA 71 USD
TCC2034-25G
Cyclohexylurea ≥98.0% (by total nitrogen basis)
Cyclohexylurea

CAS Number: 698-90-8
MDL Number: MFCD00014286
Molecular Formula: C7H14N2O
Molecular Weight: 142.20
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 185

Formula: C₇H₁₄N₂O
MW: 142.20 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00014286
CAS Number: 698-90-8
https://us.vwr.com/stibo/web/std.lang.all/90/99/28939099.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR