α,2-Dichloro-4-fluorotoluene ≥98.0%
Synonyms:
2-Chloro-1-(chloromethyl)-4-fluorobenzene
, 2-Chloro-4-fluorobenzyl chloride
C1998-25G
C1998-5G
TCC1998-25GEA
80.58
USD
TCC1998-25G
TCC1998-5G
α,2-Dichloro-4-fluorotoluene ≥98.0%
α,2-Dichloro-4-fluorotoluene
CAS Number: 93286-22-7
Molecular Formula: C7H5Cl2F
Molecular Weight: 179.02
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.37
Formula:
C₇H₅Cl₂F MW: 179.02 g/mol Density: 1.37 (20 °C) |
MDL Number:
MFCD00236026 CAS Number: 93286-22-7 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...