4-Chloro-2,5-dimethoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,5-Dimethoxy-4-chloroaniline
C1996-25G
TCC1996-25GEA
27
USD
TCC1996-25G
4-Chloro-2,5-dimethoxyaniline ≥98.0% (by GC, titration analysis)
4-Chloro-2,5-dimethoxyaniline
CAS Number: 6358-64-1
Molecular Formula: C8H10ClNO2
Molecular Weight: 187.62
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 119
Flash Point (°C): 163
Formula:
C₈H₁₀ClNO₂ MW: 187.63 g/mol Melting Pt: 119 °C Flash Pt: 163 °C |
MDL Number:
MFCD00014893 CAS Number: 6358-64-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...