Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Acetylbenzonitrile ≥98.0%

Supplier: TCI America


C1993-5G C1993-25G
TCC1993-5GEA 24.97 USD InStock
TCC1993-5G TCC1993-25G
3-Acetylbenzonitrile ≥98.0%
3-Acetylbenzonitrile
CAS Number: 6136-68-1
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 99

Other Product Names / Synonyms: 3-Acetylbenzonitrile


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 120 °C
Melting Pt: 99 °C
MDL Number: MFCD00001806
CAS Number: 6136-68-1
EINECS: 228-110-6
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty