Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlor-1,2-benzisothiazol ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloro-1,2-benzisothiazole

C1991-25G
TCC1991-25GEA 118.6 USD InStock
TCC1991-25G
3-Chlor-1,2-benzisothiazol ≥98.0%
3-Chlor-1,2-benzisothiazol
CAS Number: 7716-66-7
Molecular Formula: C7H4ClNS
Molecular Weight: 169.63
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 41


Formula: C₇H₄ClNS
MW: 169,63 g/mol
Melting Pt: 41 °C
MDL Number: MFCD00673254
CAS Number: 7716-66-7
EINECS: 000-000-0
UN: 0000
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty