1-(3-Chloropropoxy)-4-fluorobenzene ≥97.0%
C1969-25G
TCC1969-25GEA
28.86
USD
TCC1969-25G
1-(3-Chloropropoxy)-4-fluorobenzene ≥97.0%
1-(3-Chloropropoxy)-4-fluorobenzene
CAS Number: 1716-42-3
Molecular Formula: C9H10ClFO
Molecular Weight: 188.63
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 133
Specific Gravity (20/20): 1.20
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(4-Fluorophenoxy)propyl Chloride
Formula:
C₉H₁₀ClFO Boiling Pt: 133 °C Density: 1.20 (20 °C) |
MDL Number:
MFCD00018986 CAS Number: 1716-42-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...