Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyclopentene-1-carboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: cyclopent-3-ene-1-carboxylic acid

C1914-5G
TCC1914-5G
3-Cyclopentene-1-carboxylic acid ≥97.0% (by GC, titration analysis)
CAS Number: 7686-77-3
MDL Number: MFCD00101764
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 215
Specific Gravity (20/20): 1.10


Formula: C₆H₈O₂
MW: 112,13
Boiling Pt: 215 °C
Density: 1,10 (20 °C)
MDL Number: MFCD00101764
CAS Number: 7686-77-3
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)
Boiling point (°C) 215
Specific Gravity (20/20) 1.10


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty