6-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C1780-5G
C1780-1G
TCC1780-5GEA
211.15
USD
TCC1780-5G
TCC1780-1G
6-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)
6-Chloro-m-toluic acid
CAS Number: 6342-60-5
MDL Number: MFCD00045798
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 150
Other Product Names / Synonyms: 6-Chloro-m-toluic Acid
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00045798 CAS Number: 6342-60-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...