Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloromethyl polystyrene resin 1,8 - 2,5 mmol/g, cross-linked with 2% DVB (100-200mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymerMerrifield Resin

C1747-5G C1747-25G
TCC1747-5GEA 33.29 USD InStock
TCC1747-5G TCC1747-25G
Chloromethyl polystyrene resin 1,8 - 2,5 mmol/g, cross-linked with 2% DVB (100-200mesh)
Chloromethyl polystyrene resin
MDL Number: MFCD00146406
Form: Crystal
Color: White

Other Product Names / Synonyms: Merrifield Resin


MDL Number: MFCD00146406
CAS Number: 55844-94-5
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty