Chloromethyl polystyrene resin, cross-linked with 1% DVB (100-200mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymerMerrifield Resin

C1744-5G C1744-25G
TCC1744-5GEA 56.1 USD
TCC1744-5G TCC1744-25G
Chloromethyl polystyrene resin, cross-linked with 1% DVB (100-200mesh)
Chloromethyl polystyrene resin

MDL Number: MFCD00146406


Other Product Names / Synonyms: Merrifield Resin

MDL Number: MFCD00146406
CAS Number: 55844-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR