Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl (R)-4-Chloro-3-hydroxybutyrate ≥97.0%

Supplier: TCI America
Synonyms: (R)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester

C1733-25G C1733-5G
TCC1733-25G TCC1733-5G
Ethyl (R)-4-Chloro-3-hydroxybutyrate ≥97.0%
CAS Number: 90866-33-4
MDL Number: MFCD00211242
Molecular Formula: C6H11ClO3
Molecular Weight: 166.60
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: 14 deg (neat)

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester


Formula: C₆H₁₁ClO₃
MW: 166,60 g/mol
Boiling Pt: 95 °C
Density: 1,19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00211242
CAS Number: 90866-33-4
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)
Boiling point (°C) 95
Flash Point (°C) 110
Specific Gravity (20/20) 1.19
Specific rotation [a]20/D 14 deg (neat)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty