Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloronicotinamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America


C1725-5G
TCC1725-5GEA 106.09 USD InStock
TCC1725-5G
6-Chloronicotinamide ≥98.0% (by total nitrogen basis)
6-Chloronicotinamide
CAS Number: 6271-78-9
MDL Number: MFCD00006242
Molecular Formula: C6H5ClN2O
Molecular Weight: 156.57
Purity/Analysis Method: 98.0% (N)
Form: Crystal

Other Product Names / Synonyms: 6-Chloropyridine-3-carboxamide


Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD00006242
CAS Number: 6271-78-9
EINECS: 228-456-8
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (N)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty