4-Chloro-2-methylanisole ≥97.0%

Supplier: TCI America

Synonyms: 1-Chloro-4-methoxy-3-methylbenzene5-Chloro-2-methoxytoluene4-Chloro-1-methoxy-2-methylbenzene

C1724-5G
TCC1724-5GEA 43.63 USD
TCC1724-5G
4-Chloro-2-methylanisole ≥97.0%
4-Chloro-2-methylanisole

CAS Number: 3260-85-3
MDL Number: MFCD00070771
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Flash Point (°C): 101


Other Product Names / Synonyms: 4-Chloro-2-methylanisole

Formula: C₈H₉ClO
MW: 156.61 g/mol
Flash Pt: 101 °C
MDL Number: MFCD00070771
CAS Number: 3260-85-3
https://us.vwr.com/stibo/web/std.lang.all/89/24/28938924.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR