2-Chlorobenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Chloro-1H-benzimidazole

C1719-5G
TCC1719-5GEA 51.77 USD
TCC1719-5G
2-Chlorobenzimidazole ≥98.0% (by GC, titration analysis)
2-Chlorobenzimidazole

CAS Number: 4857-06-1
MDL Number: MFCD00051944
Molecular Formula: C7H5ClN2
Molecular Weight: 152.58
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00051944
CAS Number: 4857-06-1
EINECS: 225-453-3
UN: 0000

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR