5-tert-Butyl-N-carbobenzoxy-L-glutamate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-tert-Butyl N-Cbz-L-glutamateN-Cbz-L-glutamic Acid 5-tert-Butyl EsterZ-Glu(OtBu)-OHN-Carbobenzoxy-L-glutamic Acid 5-tert-Butyl Ester

C1715-5G
TCC1715-5GEA 77.14 USD
TCC1715-5G
5-tert-Butyl-N-carbobenzoxy-L-glutamate ≥98.0% (by HPLC, titration analysis)
5-tert-Butyl-N-carbobenzoxy-L-glutamate

CAS Number: 3886-08-6
MDL Number: MFCD00038274
Molecular Formula: C17H23NO6
Molecular Weight: 337.37
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 87
Specific rotation [a]20/D: -13 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Carbobenzoxy-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Cbz-L-glutamate, N-Cbz-L-glutamic Acid 5-tert-Butyl Ester, Z-Glu(OtBu)-OH

Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00038274
CAS Number: 3886-08-6
https://us.vwr.com/stibo/web/std.lang.all/89/18/28938918.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR