4-Chloro-3-nitrocoumarin ≥96.0%
Supplier: TCI America
C1691-5G
TCC1691-5GEA
129
USD
TCC1691-5G
4-Chloro-3-nitrocoumarin ≥96.0%
4-Chloro-3-nitrocoumarin
CAS Number: 38464-20-9
MDL Number: MFCD00051670
Molecular Formula: C9H4ClNO4
Molecular Weight: 225.58
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 162
Formula:
C₉H₄ClNO₄ Melting Pt: 162 °C |
MDL Number:
MFCD00051670 CAS Number: 38464-20-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...