Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-3-phenyl-2,1-benzisoxazole ≥96.0%

Supplier: TCI America


C1687-25G
TCC1687-25GEA 33.29 USD InStock
TCC1687-25G
5-Chloro-3-phenyl-2,1-benzisoxazole ≥96.0%
5-Chloro-3-phenyl-2,1-benzisoxazole
CAS Number: 719-64-2
MDL Number: MFCD00014573
Molecular Formula: C13H8ClNO
Molecular Weight: 229.66
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 115

Other Product Names / Synonyms: 5-Chloro-3-phenylanthranil


Formula: C₁₃H₈ClNO
MW: 229,67
Melting Pt: 115 °C
MDL Number: MFCD00014573
CAS Number: 719-64-2
EINECS: 211-950-2
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty