4-Chlorophenyl cyclopropyl ketone ≥98.0%
Supplier: TCI America
Synonyms:
(4-Chlorophenyl)(cyclopropyl)methanone
C1681-25G
TCC1681-25GEA
123
USD
TCC1681-25G
4-Chlorophenyl cyclopropyl ketone ≥98.0%
4-Chlorophenyl cyclopropyl ketone
CAS Number: 6640-25-1
MDL Number: MFCD00001295
Molecular Formula: C10H9ClO
Molecular Weight: 180.63
Purity/Analysis Method:
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 25
Other Product Names / Synonyms: 4-Chlorobenzoylcyclopropane
Formula:
C₁₀H₉ClO MW: 180.63 g/mol Flash Pt: 113 °C |
MDL Number:
MFCD00001295 CAS Number: 6640-25-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...