2-Chloro-4-fluoroanisole ≥97.0%
Supplier: TCI America
C1667-5G
TCC1667-5GEA
106.58
USD
TCC1667-5G
2-Chloro-4-fluoroanisole ≥97.0%
2-Chloro-4-fluoroanisole
CAS Number: 2267-25-6
MDL Number: MFCD00672965
Molecular Formula: C7H6ClFO
Molecular Weight: 160.57
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 77
Specific Gravity (20/20): 1.30
Formula:
C₇H₆ClFO MW: 160.58 g/mol Boiling Pt: 200 °C Density: 1.30 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00672965 CAS Number: 2267-25-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...