2-Chloroethanesulfonic acid sodium salt hydrate ≥95.0% (by titrimetric analysis)
Synonyms:
Sodium-2-chloroethanesulfonate
, Sodium-2-chloroethanesulphonate
C1656-25G
TCC1656-025GEA
150.96
USD
TCC1656-025G
2-Chloroethanesulfonic acid sodium salt hydrate ≥95.0% (by titrimetric analysis)
2-Chloroethanesulfonic acid sodium salt
CAS Number: 15484-44-3
MDL Number: MFCD00007531
Molecular Formula: C2H5ClO3S
Molecular Weight: 166.55
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2-Chloroethanesulfonic Acid Sodium Salt
Formula:
C₂H₄ClNaO₃S MW: 166.55 g/mol |
MDL Number:
MFCD00007531 CAS Number: 15484-44-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...