Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chromanol ≥98.0%

Supplier: TCI America


C1627-1G
TCC1627-001GEA 45.77 USD InStock
TCC1627-001G
4-Chromanol ≥98.0%
4-Chromanol
CAS Number: 1481-93-2
MDL Number: MFCD00006849
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 80
Melting point (°C): 43
Flash Point (°C): 113

Other Product Names / Synonyms: 3,4-Dihydro-2H-1-benzopyran-4-ol, 4-Hydroxychroman


Formula: C₉H₁₀O₂
MW: 150,18
Boiling Pt: 80 °C
Melting Pt: 43 °C
Flash Pt: 113 °C
MDL Number: MFCD00006849
CAS Number: 1481-93-2
EINECS: 216-041-4
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty