Ethyl-4-chlorobenzoate ≥96.0%
Supplier: TCI America
Synonyms:
Ethyl 4-chlorobenzoate
, 4-Chlorobenzoic acid ethyl ester
C1624-25G
TCC1624-25GEA
106.58
USD
TCC1624-25G
Ethyl-4-chlorobenzoate ≥96.0%
Ethyl 4-chlorobenzoate
CAS Number: 7335-27-5
MDL Number: MFCD00013645
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 238
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 4-Chlorobenzoic Acid Ethyl Ester
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol Boiling Pt: 238 °C Density: 1.18 (20 °C) |
MDL Number:
MFCD00013645 CAS Number: 7335-27-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...