N-Cbz-L-phenylalaninol ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
N-Benzyloxycarbonyl-L-phenylalaninol
, (S)-2-(Cbz-amino)-3-phenyl-1-propanol
, Z-Phenylalaninol
CAS Number: 6372-14-1
MDL Number: MFCD00191138
Molecular Formula: C17H19NO3
Molecular Weight: 285.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: -42 deg (C=2, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-N-Carbobenzoxy-2-amino-3-phenyl-1-propanol, (S)-N-Cbz-2-amino-3-phenyl-1-propanol, N-Cbz-L-phenylalaninol
Formula:
C₁₇H₁₉NO₃ MW: 285.3 g/mol Melting Pt: 93 °C |
MDL Number:
MFCD00191138 CAS Number: 6372-14-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...