4-Cyano-4'-heptyloxybiphenyl ≥98.0%

Supplier: TCI America

C1606-1G
TCC1606-001GEA 56.21 USD
TCC1606-001G
4-Cyano-4'-heptyloxybiphenyl ≥98.0%
4-Cyano-4'-heptyloxybiphenyl

CAS Number: 52364-72-4
MDL Number: MFCD00134146
Molecular Formula: C20H23NO
Molecular Weight: 293.41
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 54


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-Heptyloxy-4-biphenylcarbonitrile, 7OCB

Formula: C₂₀H₂₃NO
Melting Pt: 54 °C
MDL Number: MFCD00134146
CAS Number: 52364-72-4
https://us.vwr.com/stibo/web/std.lang.all/88/23/28938823.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR