Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentadienyliron Dicarbonyl ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Cyclopentadienyliron dicarbonyl dimerBis(cyclopentadienyl)tetracarbonyl Diiron

C1592-5G
TCC1592-005G
Cyclopentadienyliron Dicarbonyl ≥96.0% (by titrimetric analysis)
CAS Number: 12154-95-9
MDL Number: MFCD00001436
Molecular Formula: C14H10Fe2O4
Molecular Weight: 353.92
Purity/Analysis Method: 96.0% (T)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Bis(cyclopentadienyl)tetracarbonyl Diiron


Formula: C₁₄H₁₀Fe₂O₄
MW: 353,92 g/mol
MDL Number: MFCD00001436
CAS Number: 12154-95-9
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)
Color Deep Yellow Red


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty