Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0%

Supplier: TCI America

Synonyms: 1,4-Cyclohexanedione bisethylene ketal1,4-Cyclohexanedione bis(ethylene acetal)

C1544-25G
TCC1544-025GEA 72.83 USD InStock
TCC1544-025G
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0%
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan
CAS Number: 183-97-1
MDL Number: MFCD00010851
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 78

Other Product Names / Synonyms: 1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecane


Formula: C₁₀H₁₆O₄
MW: 200,23
Melting Pt: 78 °C
MDL Number: MFCD00010851
CAS Number: 183-97-1
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty