Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorodiphenyl sulphone ≥96.0%

Supplier: TCI America


C1451-25G
TCC1451-025GEA 70.72 USD InStock
TCC1451-025G
4-Chlorodiphenyl sulphone ≥96.0%
4-Chlorodiphenyl sulfone
CAS Number: 80-00-2
MDL Number: MFCD00007549
Molecular Formula: C12H9ClO2S
Molecular Weight: 252.71
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Boiling point (°C): 389
Melting point (°C): 92

Other Product Names / Synonyms: 4-Chlorodiphenyl Sulfone, 1-Chloro-4-(phenylsulfonyl)benzene, Sulferone


Formula: C₁₂H₉ClO₂S
Boiling Pt: 389 °C
Melting Pt: 92 °C
MDL Number: MFCD00007549
CAS Number: 80-00-2
EINECS: 201-242-1
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty