2-Cyano-4'-methylbiphenyl ≥99.0%

Supplier: TCI America

Synonyms: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

C1446-10G
TCC1446-010GEA 37 USD
TCC1446-010G
2-Cyano-4'-methylbiphenyl ≥99.0%
2-Cyano-4'-methylbiphenyl

CAS Number: 114772-53-1
MDL Number: MFCD00151805
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 100


Other Product Names / Synonyms: 4'-Methyl-2-biphenylcarbonitrile

Formula: C₁₄H₁₁N
MW: 193.26 g/mol
Melting Pt: 53 °C
Flash Pt: 100 °C
MDL Number: MFCD00151805
CAS Number: 114772-53-1
https://us.vwr.com/stibo/web/std.lang.all/86/71/28938671.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR