2-Chloroethyl phenyl sulfide ≥98.0%
Supplier: TCI America
C1388-25G
C1388-5G
TCC1388-025GEA
115.71
USD
TCC1388-025G
TCC1388-005G
2-Chloroethyl phenyl sulfide ≥98.0%
2-Chloroethyl phenyl sulphide
CAS Number: 5535-49-9
MDL Number: MFCD00013691
Molecular Formula: C8H9ClS
Molecular Weight: 172.67
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 104
Specific Gravity (20/20): 1.18
Formula:
C₈H₉ClS MW: 172.68 g/mol Boiling Pt: 245 °C Density: 1.18 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00013691 CAS Number: 5535-49-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...