TCIAC1284

C1284-10G
TCC1284-010GEA 106.87 USD
TCC1284-010G
TCIAC1284
CAS Number: 53233-89-9
MDL Number: MFCD00023474
Molecular Formula: C5H4ClNO2
Molecular Weight: 145.54
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 295

Other Product Names / Synonyms: 5-Chloro-2,3-dihydroxypyridine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR