TCIAC1260

C1260-25G
TCC1260-25GEA 45.64 USD
TCC1260-25G
TCIAC1260
CAS Number: 32832-01-2
MDL Number: MFCD00065268
Molecular Formula: C34H48Cl2O2
Molecular Weight: 559.66
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 135

Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: Cholesteryl 2,4-Dichlorobenzoate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR