TCIAC1236

C1236-5G
TCC1236-005GEA 72.86 USD
TCC1236-005G
TCIAC1236
CAS Number: 1851-09-8
MDL Number: MFCD00045626
Molecular Formula: C8H6ClNO2S
Molecular Weight: 215.65
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 170

Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: 4-Chlorophenylsulfonylacetonitrile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR