Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(3-Chlorophenoxy)propionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America


C1094-25G
TCC1094-025GEA 37.45 USD InStock
TCC1094-025G
2-(3-Chlorophenoxy)propionic acid ≥98.0% (by titrimetric analysis)
2-(3-Chlorophenoxy)propionic acid
CAS Number: 101-10-0
MDL Number: MFCD00009723
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 114


Formula: C₉H₉ClO₃
Melting Pt: 114 °C
MDL Number: MFCD00009723
CAS Number: 101-10-0
EINECS: 202-915-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty