Carbimazole ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
Ethyl 3-methyl-2-thioxo-2,3-dihydro-1H-imidazole-1-carboxylate
, 1-Ethoxycarbonyl-3-methyl-2-thioxo-4-imidazoline
C1080-25G
C1080-5G
TCC1080-5GEA
65.98
USD
TCC1080-5G
TCC1080-025G
Carbimazole ≥95.0% (by HPLC)
Carbimazole
CAS Number: 22232-54-8
MDL Number: MFCD00027421
Molecular Formula: C7H10N2O2S
Molecular Weight: 186.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 124
Other Product Names / Synonyms: 1-Ethoxycarbonyl-3-methyl-2-thioxo-4-imidazoline
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol Melting Pt: 124 °C |
MDL Number:
MFCD00027421 CAS Number: 22232-54-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...