9-Cyanoanthracene ≥95.0%

Supplier: TCI America

Synonyms: 9-Anthracenecarbonitrile

C1070-25G C1070-5G C1070-1G
TCC1070-25GEA 137.4 USD
TCC1070-25G TCC1070-005G TCC1070-001G
9-Cyanoanthracene ≥95.0%
9-Cyanoanthracene

CAS Number: 1210-12-4
MDL Number: MFCD00001242
Molecular Formula: C15H9N
Molecular Weight: 203.24
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 175


Other Product Names / Synonyms: 9-Anthracenecarbonitrile

Formula: C₁₅H₉N
Melting Pt: 175 °C
MDL Number: MFCD00001242
CAS Number: 1210-12-4
https://us.vwr.com/stibo/web/std.lang.all/83/33/28938333.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR